Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals
Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals
Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals
Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals
Suitable for use as a polarographic supporting electrolyte
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF043999.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS42753-71-9
IUPAC Name6-amino-3-bromo-2-methylpyridin-1-ium
Molecular FormulaC6H8BrN2
InChI KeySEOZHXRTVJPQPZ-UHFFFAOYSA-O
SMILESCC1=[NH+]C(N)=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (GC)≥96.0%
Melting Point (clear melt)76.5-85.5?C
Appearance (Color)White to pale cream
Suitable for use as a polarographic supporting electrolyteTetra-n-butylammonium perchlorate is used as a supporting electrolyte in polarographic measurements.