Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals
Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals
Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals
Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetra-n-butylammonium perchlorate, electrochemical grade, Thermo Scientific Chemicals
Suitable for use as a polarographic supporting electrolyte
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number ALF043999.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTetra-n-butylammonium perchlorate
CAS1923-70-2
Health Hazard 1H272-H315-H319-H335
Health Hazard 2GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
Suitable for use as a polarographic supporting electrolyteTetra-n-butylammonium perchlorate is used as a supporting electrolyte in polarographic measurements.