Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
As spectroscopic standard, cobalt plating, catalysis, ceramics
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF043902.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium cobalt (II) sulfate hexahydrate
Melting Point∼120°C (decomposition)
CAS13586-38-4
ColorPink to Red
Health Hazard 1H302-H317-H350
View more
Ammonium cobalt(II) sulfate hexahydrate is uses as spectroscopic standard, cobalt plating, catalysis and ceramics. It is also used in pharmaceutical intermediates.