(2-Hydroxyethyl)hydrazine, Thermo Scientific Chemicals
(2-Hydroxyethyl)hydrazine, Thermo Scientific Chemicals
(2-Hydroxyethyl)hydrazine, Thermo Scientific Chemicals
(2-Hydroxyethyl)hydrazine, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(2-Hydroxyethyl)hydrazine, Thermo Scientific Chemicals
CAS: 109-84-2 | C2H8N2O | 76.099 g/mol
Have Questions?
Catalog NumberQuantity
ALF043793.1850 g
Catalog number ALF043793.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS101-84-8
IUPAC Namephenoxybenzene
Molecular FormulaC12H10O
InChI KeyUSIUVYZYUHIAEV-UHFFFAOYSA-N
SMILESO(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.5790-1.5820 @ 20?C
(2-Hydroxyethyl)hydrazine is used as a plant growth regulant, a component in jet fuels, intermediate for organic synthesis especially heterocycles used in agrochemicals, pharmaceuticals, stabilizers and polymerizations, Chain extender for urethane formulations, corrosion inhibitor.