Aluminum wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
Aluminum wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
Aluminum wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
Aluminum wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
Aluminum wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aluminum wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 m
5 m
Catalog number ALF043427.G9
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 m
Request bulk or custom format
Chemical Identifiers
CAS2235-46-3
IUPAC NameN,N-diethyl-3-oxobutanamide
Molecular FormulaC8H15NO2
InChI KeyNTMXFHGYWJIAAE-UHFFFAOYSA-N
SMILESCCN(CC)C(=O)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Assay (GC)≥96.0%
Refractive Index1.4695-1.4745 @ 20?C