2-Propanethiol, 98%, Thermo Scientific Chemicals
2-Propanethiol, 98%, Thermo Scientific Chemicals
2-Propanethiol, 98%, Thermo Scientific Chemicals
2-Propanethiol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Propanethiol, 98%, Thermo Scientific Chemicals
CAS: 75-33-2 | C3H8S | 76.157 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF043422.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS461-96-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4970-1.5010 @ 20?C
2-Propanethiol Reacts with 3-chloro-propene to get 3-isopropylsulfanyl-propene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only