1-Propanethiol, 98%, Thermo Scientific Chemicals
1-Propanethiol, 98%, Thermo Scientific Chemicals
1-Propanethiol, 98%, Thermo Scientific Chemicals
1-Propanethiol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Propanethiol, 98%, Thermo Scientific Chemicals
CAS: 107-03-9 | C3H8S | 76.157 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF043420.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS342573-75-5
IUPAC Name1-ethyl-3-methyl-1H-imidazol-3-ium ethyl sulfate
Molecular FormulaC8H16N2O4S
InChI KeyVRFOKYHDLYBVAL-UHFFFAOYSA-M
SMILESCCOS([O-])(=O)=O.CCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Ion Chromatography≥98.5%
Water Content (Karl Fischer Titration)≤1,500 ppm
Proton NMRConforms to structure
Refractive Index1.4780-1.4810 @ 20?C
View more
1-Propanethiol, is used widely in research areas. This compound has a wide range of applications including cell biology, materials science, nanoscience, self-assembled monolayers (SAMs), nanoelectronics, and contact printing. It is also used as a chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only