Nickel powder, -325 mesh, typically 99.8% (metals basis), Thermo Scientific Chemicals
Nickel powder, -325 mesh, typically 99.8% (metals basis), Thermo Scientific Chemicals
Nickel powder, -325 mesh, typically 99.8% (metals basis), Thermo Scientific Chemicals
Nickel powder, -325 mesh, typically 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nickel powder, -325 mesh, typically 99.8% (metals basis), Thermo Scientific Chemicals
Form: Conductive dendritic powder
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF043214.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNickel
ColorSilver to Gray
CAS7440-02-0
Health Hazard 1H228-H317-H350-H372
Health Hazard 2Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.
View more
Conductive surface coatings, inks, adhesives, and sealants