Ammonium hexafluorotitanate, 99.99% (metals basis), Thermo Scientific Chemicals
Ammonium hexafluorotitanate, 99.99% (metals basis), Thermo Scientific Chemicals
Ammonium hexafluorotitanate, 99.99% (metals basis), Thermo Scientific Chemicals
Ammonium hexafluorotitanate, 99.99% (metals basis), Thermo Scientific Chemicals
Ammonium hexafluorotitanate, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hexafluorotitanate, 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 16962-40-6 | (NH4)2TiF6
Have Questions?
Catalog NumberQuantity
ALF043192.1425 g
Catalog number ALF043192.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium hexafluorotitanate
CAS16962-40-6
ColorWhite
Health Hazard 1H302+H312+H332
Health Hazard 2GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
View more
Ammonium hexafluorotitanate is used as an anti-corrosive cleaning agent. It is also used for the production of ceramics and glass. Further, it is used in the preparation of synthetic gems.