Methyl cellulose, viscosity 400 cPs
Methyl cellulose, viscosity 400 cPs
Methyl cellulose, viscosity 400 cPs
Methyl cellulose, viscosity 400 cPs
Thermo Scientific Chemicals
Methyl cellulose, viscosity 400 cPs
CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF043147.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS6212-33-5
IUPAC Name(2R)-2-azaniumyl-2-(4-chlorophenyl)acetate
Molecular FormulaC8H8ClNO2
InChI KeyQGJGBYXRJVIYGA-SSDOTTSWSA-N
SMILES[NH3+][C@@H](C([O-])=O)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
Methyl cellulose is used as a thickener and emulsifier in various food and cosmetic products like hair shampoos, toothpastes and liquid soaps. It is used in the preparation of capsules in nutritional supplements. It is also used as a gel in advanced cookery and as a lubricant. It is also involved in the treatment of constipation. It acts as a buffer additive in capillary electrophoresis to control electoosmotic flow for improved separations. Further, it is used as a performance additive in construction materials and as sizing in the production of papers and textiles.