Aluminum dihydrogen phosphate, 50% w/w aq. soln.
Aluminum dihydrogen phosphate, 50% w/w aq. soln.
Actual product may vary
Thermo Scientific Chemicals
Aluminum dihydrogen phosphate, 50% w/w aq. soln.
CAS: 13530-50-2 | AlH{6}O{1}{2}P{3}
Have Questions?
Catalog NumberQuantity
ALF042858.A75 kg
Catalog number ALF042858.A7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Aluminum dihydrogen phosphate is used as intermediates and solid separation agents. It is also used in petroleum, chemical industry, shipbuilding, space technology, electric industry, high-temperature kilns, heat treatment resistance furnaces and electric insulation. Further, it is used as inorganic coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS13530-50-2
Physical FormLiquid
Quantity5 kg
Molecular FormulaAlH{6}O{1}{2}P{3}
SynonymAluminum hydrogen phosphate;Aluminum phosphate monobasic
Unit Size