Silica gel 60, 0.075-0.2mm (70-200 mesh)
Silica gel 60, 0.075-0.2mm (70-200 mesh)
Silica gel 60, 0.075-0.2mm (70-200 mesh)
Have Questions?
Change viewbuttonViewtableView
Particle Size:
70/150 Mesh
-70/200 Mesh
Quantity:
View all
1 kg
Surface Area (Metric):
View all
500 to 600 m2/g
Catalog number ALF042723.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Particle Size:
70/150 Mesh
Quantity:
1 kg
Surface Area (Metric):
500 to 600 m2/g
Request bulk or custom format
Chemical Identifiers
CAS70-18-8
IUPAC Name2-amino-4-({1-[(carboxymethyl)carbamoyl]-2-sulfanylethyl}carbamoyl)butanoic acid
Molecular FormulaC10H17N3O6S
InChI KeyRWSXRVCMGQZWBV-UHFFFAOYNA-N
SMILESNC(CCC(=O)NC(CS)C(=O)NCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Iron (Fe)=<10 ppm
Appearance (Color)White to almost white
Infrared spectrumConforms
Titration Iodimetric97.5 to 102.5 %
View more
Classic column chromatography with no applied pressure (gravity pulled)Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.