Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals
Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals
Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals
Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals
CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALF042679.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS12024-21-4
IUPAC Namedigallium(3+) trioxidandiide
Molecular FormulaGa2O3
InChI KeyAJNVQOSZGJRYEI-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Ga+3].[Ga+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total trace metal impurities=<0.010 %
Appearance (Form)Crystalline powder
Potassium chloride 0.010M solution is used as conductive standard and as electrolyte in analytical chemistry.