Copper powder, spherical, -100+325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Copper powder, spherical, -100+325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Copper powder, spherical, -100+325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Copper powder, spherical, -100+325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper powder, spherical, -100+325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Oxygen analysis included
Have Questions?
Catalog NumberQuantity
ALF042623.36500 g
Catalog number ALF042623.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS638-16-4
IUPAC Name1,3,5-triazinane-2,4,6-trithione
Molecular FormulaC3H3N3S3
InChI KeyWZRRRFSJFQTGGB-UHFFFAOYSA-N
SMILESS=C1NC(=S)NC(=S)N1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=94.0 %
Appearance (Color)Yellow
Infrared spectrumConforms
Appearance (Form)Powder