Silver(I) oxide, 99.99% (metals basis), Thermo Scientific™
Silver(I) oxide, 99.99% (metals basis), Thermo Scientific™
Silver(I) oxide, 99.99% (metals basis), Thermo Scientific™
Silver(I) oxide, 99.99% (metals basis), Thermo Scientific™
Thermo Scientific Chemicals
Silver(I) oxide, 99.99% (metals basis), Thermo Scientific™
Have Questions?
Catalog number ALF042577.49
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver(I) oxide
Melting Point230°C (decomposition)
CAS20667-12-3
ColorBrown
Health Hazard 1H271-H318
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.