7-Methylquinoline, 97%, Thermo Scientific Chemicals
7-Methylquinoline, 97%, Thermo Scientific Chemicals
7-Methylquinoline, 97%, Thermo Scientific Chemicals
7-Methylquinoline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
7-Methylquinoline, 97%, Thermo Scientific Chemicals
CAS: 612-60-2 | C10H9N | 143.189 g/mol
Have Questions?
Catalog NumberQuantity
ALF042475.031 g
Catalog number ALF042475.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS13035-19-3
IUPAC Namepiperidin-4-amine
Molecular FormulaC5H12N2
InChI KeyBCIIMDOZSUCSEN-UHFFFAOYSA-N
SMILESNC1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange to brown
FormLiquid or low melting solid
Assay (GC)≥94.0%
7-methylquinoline has been bioassayed as tumor initiator on the skin of Sencar mice.