ORP test solution, 200-275 mV, Thermo Scientific Chemicals
ORP test solution, 200-275 mV, Thermo Scientific Chemicals
Thermo Scientific Chemicals
ORP test solution, 200-275 mV, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
250 mL
Catalog number ALF042131.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS28169-46-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
Assay (Aqueous acid-base Titration)≥97.0 to ≤103.0%
Melting Point (clear melt)204-210°C
FormCrystals or crystalline powder or powder
Assay (Silylated GC)≥97.0%
Test solution for ORP electrodes