Sodium oxalate, ACS, 99.5+%, Thermo Scientific Chemicals
Sodium oxalate, ACS, 99.5+%, Thermo Scientific Chemicals
Sodium oxalate, ACS, 99.5+%, Thermo Scientific Chemicals
Sodium oxalate, ACS, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium oxalate, ACS, 99.5+%, Thermo Scientific Chemicals
CAS: 62-76-0 | C2Na2O4 | 134.00 g/mol
Have Questions?
Catalog NumberQuantity
ALF041759.A11 kg
Catalog number ALF041759.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS1468-83-3
IUPAC Name1-(thiophen-3-yl)ethan-1-one
Molecular FormulaC6H6OS
InChI KeyRNIDWJDZNNVFDY-UHFFFAOYSA-N
SMILESCC(=O)C1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)≥97.5%
Melting Point (clear melt)57.0-63.0?C
FormCrystals or powder or crystalline powder
Sodium oxalate is used as a reducing agent, as a primary standard for standardizing potassium permanganate solutions and as a neutralizer for exothermic acidic reactions. It finds application in metal cleaning preparations, metal extraction and separation, blueprint coatings, electroplating baths, special cement manufacturing, plant nutrition as well as in textile and pharmaceutical industries. It plays an important role to remove calcium ions from blood plasma and prevents blood clotting.