Humic acid sodium salt, tech. 50-60% (as humic acid), Thermo Scientific Chemicals
Humic acid sodium salt, tech. 50-60% (as humic acid), Thermo Scientific Chemicals
Humic acid sodium salt, tech. 50-60% (as humic acid), Thermo Scientific Chemicals
Humic acid sodium salt, tech. 50-60% (as humic acid), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Humic acid sodium salt, tech. 50-60% (as humic acid), Thermo Scientific Chemicals
CAS: 68131-04-4 | C9H8Na2O4 | 226.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Catalog number ALF041748.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHumic acid sodium salt
CAS68131-04-4
Melting Point>300°C
Health Hazard 1H315-H319-H335-H500
ColorBlack
View more
Humic acid sodium salt is a principal component of humic substances. It boosts plant growth.