Humic acid, Thermo Scientific Chemicals
Humic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Humic acid, Thermo Scientific Chemicals
CAS: 1415-93-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALF041747.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS56-86-0
IUPAC Name2-aminopentanedioic acid
Molecular FormulaC5H9NO4
InChI KeyWHUUTDBJXJRKMK-UHFFFAOYNA-N
SMILESNC(CCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥98.5%
Identification (FTIR)Conforms
Optical Rotation+32?? 2? (c= 5 in 5N HCl)
Appearance (Color)White
FormCrystals or powder or crystalline powder
Metal chelator; pigments.Humic acid is used as a soil supplement in agriculture and human nutritional supplement. It is used to improve the growth and cultivation of crops, citrus, turf, flowers. It is also used to improve the strength of organically-deficient soils. It is utilized to stimulate the immune system and treating the influenza, avian flu, swine flu and other viral infections.