Cobalt(II) phthalocyanine, Thermo Scientific Chemicals
Cobalt(II) phthalocyanine, Thermo Scientific Chemicals
Cobalt(II) phthalocyanine, Thermo Scientific Chemicals
Cobalt(II) phthalocyanine, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cobalt(II) phthalocyanine, Thermo Scientific Chemicals
CAS: 3317-67-7 | C32H16CoN8 | 571.469 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number ALF041496.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS4229-91-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale green to yellow
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.4410-1.4440 @ 20?C
Cobalt(II) phthalocyanine may be used to develop carbon black based electrocatalysts. CoPC may be used as an end capping agent of a hyperbranched poly(aryl ether ketone) to be used for oxidative decomposition of 2,4,6,-trichlorophenol. CoPC modified carbon paste may be used as indicators for electrocatalytic amperometric measurements. ZnO impregnated with CoPC may be used as a sensitizer to determine the photodegradation of cyanide in aqueous suspension. CoPC catalyzed aerobic regenerations of aldehydes and ketones have been investigated.