Nafion∣r NR50, super acidic ion exchange resin catalyst, Thermo Scientific Chemicals
Nafion∣r NR50, super acidic ion exchange resin catalyst, Thermo Scientific Chemicals
Nafion∣r NR50, super acidic ion exchange resin catalyst, Thermo Scientific Chemicals
Nafion∣r NR50, super acidic ion exchange resin catalyst, Thermo Scientific Chemicals
Nafion∣r NR50, super acidic ion exchange resin catalyst, Thermo Scientific Chemicals
Super acid catalyst
Have Questions?
Catalog NumberQuantity
ALF040802.0910 g
Catalog number ALF040802.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS471-25-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to brown
Infrared spectrumConforms
Titration with NaOH97.5 to 102.5 %
Appearance (Form)Liquid or solid
Refractive index1.4305 to 1.4345 (20°C, 589 nm)
Super acid catalyst Nafion™ NR50 is a perfluorinated ion-exchange membrane which has a wide variety of commercial uses. It is an acidic hydrogen ion form and superacid catalysts useful in a wide variety of synthetic applications. It finds application as a catalyst in alkylation and acylation reactions. It is also used in the production of sensors.