Calcium sulfate, anhydrous, 99%, Thermo Scientific Chemicals
Calcium sulfate, anhydrous, 99%, Thermo Scientific Chemicals
Calcium sulfate, anhydrous, 99%, Thermo Scientific Chemicals
Calcium sulfate, anhydrous, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calcium sulfate, anhydrous, 99%, Thermo Scientific Chemicals
CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Catalog number ALF040144.A7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Request bulk or custom format
Chemical Identifiers
CAS118-00-3
IUPAC Name2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular FormulaC10H13N5O5
InChI KeyNYHBQMYGNKIUIF-UUOKFMHZSA-N
SMILESNC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤6%
Appearance (Color)White to pale cream
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5% (dry wt. basis)
Optical Rotation-66° to -74° (c=1.5 in 0.1N NaOH)
View more
Used as Portland-cement retarder; tile and plaster; It is the source of sulfur and sulfuric acid; Also used in polishing powder and paints, dyeing and calico printing. It is likewise employed in zinc metallurgy. It is employed in drying industrial gases, solids and many organic liquids. In granulated form it is used as soil conditioner. Used for food additives in the soy manufacturing industry.