Magnesium lactate trihydrate, 99%, Thermo Scientific Chemicals
Magnesium lactate trihydrate, 99%, Thermo Scientific Chemicals
Magnesium lactate trihydrate, 99%, Thermo Scientific Chemicals
Magnesium lactate trihydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Magnesium lactate trihydrate, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALF039369.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMagnesium lactate trihydrate
CAS18917-96-9
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362