Iron(II) phthalocyanine, 96%, Thermo Scientific Chemicals
Iron(II) phthalocyanine, 96%, Thermo Scientific Chemicals
Iron(II) phthalocyanine, 96%, Thermo Scientific Chemicals
Iron(II) phthalocyanine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iron(II) phthalocyanine, 96%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF039262.031 g
Catalog number ALF039262.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS82-57-5
IUPAC Name4-methoxy-7-methyl-5H-furo[3,2-g]chromen-5-one
Molecular FormulaC13H10O4
InChI KeyNZVQLVGOZRELTG-UHFFFAOYSA-N
SMILESCOC1=C2C=COC2=CC2=C1C(=O)C=C(C)O2
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point139°C to 142°C
GC>=96.0 %
Appearance (Color)Beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Iron phthalocyanine is been investigated as organic electroluminescence materials for the applications in organic solar cells, biosensitizers and display devices such as OLED(Organic Light Emiting Diode), OTFT(Organic Thin Film Transistor), Wearable Display, and e-paper. It is also used in the formation of new type of catalyst.