Iron(III) hexacyanoferrate(II), Thermo Scientific Chemicals
Iron(III) hexacyanoferrate(II), Thermo Scientific Chemicals
Iron(III) hexacyanoferrate(II), Thermo Scientific Chemicals
Iron(III) hexacyanoferrate(II), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iron(III) hexacyanoferrate(II), Thermo Scientific Chemicals
CAS: 14038-43-8 | C18Fe7N18 | 859.239 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF039259.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS21645-51-2
IUPAC Namealuminium(3+) trihydroxide
Molecular FormulaAlH3O3
InChI KeyWNROFYMDJYEPJX-UHFFFAOYSA-K
SMILES[OH-].[OH-].[OH-].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Chloride content≤0.85%
pH≤10.0
FormPowder, amorphous
Assay (special)Acid Neutralizing Capacity: ≥25.0meq/g
View more
Iron(III) hexacyanoferrate(II) is uses extensively as an Iron Oxide dye. Prussian blue is used as a paint and wallpaper printing, chemical coatings, carbon paper and in the plastics industry. as an antidote for poisoning with radioactive cesium or thallium. In the metalworking and mechanical engineering Prussian blue is thinly applied as a paste on metal surfaces in order to assess the quality scraped surfaces can.