Caffeine, 99.7%, Thermo Scientific Chemicals
Caffeine, 99.7%, Thermo Scientific Chemicals
Caffeine, 99.7%, Thermo Scientific Chemicals
Caffeine, 99.7%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Caffeine, 99.7%, Thermo Scientific Chemicals
CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALF039214.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS3051-09-0
IUPAC Nameammonium 2,4,6-trioxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1,3-diazinan-5-ide
Molecular FormulaC8H8N6O6
InChI KeyPCCZWUCAYILEMX-UHFFFAOYSA-O
SMILES[NH4+].O=C1NC(=O)[C-](N=C2C(=O)NC(=O)NC2=O)C(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Suitability(as complexom. indicator) Passes test
Appearance (Color)Bordeaux to red-purple
Infrared spectrumConforms
It acts as a stimulant drug and a reversible acetylcholinesterase inhibitor. It plays an important role for the inactivation of Escherichia coli O 157:H 7 in brain heart infusion (BHI) broth. It is also useful for the protection from photodamage after ultraviolet irradiation.