Ammonium copper(II) chloride dihydrate, Thermo Scientific Chemicals
Ammonium copper(II) chloride dihydrate, Thermo Scientific Chemicals
Ammonium copper(II) chloride dihydrate, Thermo Scientific Chemicals
Ammonium copper(II) chloride dihydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium copper(II) chloride dihydrate, Thermo Scientific Chemicals
CAS: 10060-13-6 | Cl4CuH12N2O2 | 277.454 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF039127.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium copper(II) chloride dihydrate
Melting Point110°C (decomposition)
CAS10060-13-6
ColorBlue
Health Hazard 1H315-H319-H335
View more
Ammonium copper(II) chloride dihydrate is used in the determination iron of content in steel. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.