1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT, Thermo Scientific Chemicals
1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT, Thermo Scientific Chemicals
1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT, Thermo Scientific Chemicals
1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT, Thermo Scientific Chemicals
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number ALF039118.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS80945-86-4
IUPAC Name6-bromo-2-chloro-1,3-benzothiazole
Molecular FormulaC7H3BrClNS
InChI KeyIJQSMNIZBBEBKI-UHFFFAOYSA-N
SMILESClC1=NC2=CC=C(Br)C=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥96.0%
Melting Point (clear melt)96.0-105.0?C
FormPowder
1,4-Dioxane is used as a stabilizer for 1,1,1-trichloroethane and chlorinated solvents. It acts as a solvent for cellulose acetate, ethyl cellulose, benzyl cellulose, resins, oils, waxes, dyes, other organic and inorganic compounds. It acts as a substitute for tetrahydrofuran due to its higher boiling point and lower toxicity. Furthermore, it is used as an internal standard for proton NMR spectroscopy in deuterium oxide. It plays an important role in liquid scintillation counting in biological procedures and as a wetting and dispersing agent in textile processing. It is involved in the preparation of dimethylmagnesium by reacting with methylmagnesium bromide and in the preparation of histological sections for microscopic examination.