Iodine monochloride, Thermo Scientific Chemicals
Iodine monochloride, Thermo Scientific Chemicals
Iodine monochloride, Thermo Scientific Chemicals
Iodine monochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodine monochloride, Thermo Scientific Chemicals
CAS: 7790-99-0 | ClI | 162.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number ALF039104.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS82257-09-8
IUPAC Name3-bromo-4-methoxypyridine
Molecular FormulaC6H6BrNO
InChI KeyDSFJKFHIMBVWAV-UHFFFAOYSA-N
SMILESCOC1=C(Br)C=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
ICl is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.