Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT, Thermo Scientific Chemicals
Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT, Thermo Scientific Chemicals
Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT, Thermo Scientific Chemicals
Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrahydrofuran, non-UV, HPLC Grade, 99.7+%, stab. with 250ppm BHT, Thermo Scientific Chemicals
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 1 L
1 L
Catalog number ALF038994.M1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 x 1 L
Request bulk or custom format
Chemical Identifiers
CAS76-54-0
IUPAC Name2',7'-dichloro-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular FormulaC20H10Cl2O5
InChI KeyVFNKZQNIXUFLBC-UHFFFAOYSA-N
SMILESOC1=CC2=C(C=C1Cl)C1(OC(=O)C3=CC=CC=C13)C1=CC(Cl)=C(O)C=C1O2
View more
SpecificationsSpecification SheetSpecification Sheet
Specific absorbance A (1%/1cm)>=2000 (at lambda max.)
Appearance (Color)Light orange to red-brown
Infrared spectrumConforms
UV Lambda max501 to 503 nm (in 0.1 N NaOH)
Appearance (Form)Powder