Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals
Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals
Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals
Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals
Air Sensitive
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Catalog number ALF036694.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS109-86-4
IUPAC Name2-methoxyethan-1-ol
Molecular FormulaC3H8O2
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 %
Refractive index1.4010 to 1.4040 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<15 APHA
Infrared spectrumConforms
View more
Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes, and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration, and dehydration, and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.