Tin granules, ACS, 99.5% min, Thermo Scientific Chemicals
Tin granules, ACS, 99.5% min, Thermo Scientific Chemicals
Tin granules, ACS, 99.5% min, Thermo Scientific Chemicals
Tin granules, ACS, 99.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tin granules, ACS, 99.5% min, Thermo Scientific Chemicals
Form: approx. 9.1g/50x50mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF036691.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTin
ColorGray
Name NoteFor analysis ACS
CAS7440-31-5
Health Hazard 1H303: May be harmful if swallowed.