Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Reagent for organic synthesis
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Catalog number ALF036609.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS70-34-8
IUPAC Name1-fluoro-2,4-dinitrobenzene
Molecular FormulaC6H3FN2O4
InChI KeyLOTKRQAVGJMPNV-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid or low melting solid
GC>=97.5 %
Appearance (Color)Yellow to brown
Infrared spectrumConforms
Refractive index1.5675 to 1.5705 (20°C, 589 nm)
Used as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It act as a catalyst for asymmetric reactions. It plays an essential role as catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.