Acrylamide, 99.9%, Thermo Scientific Chemicals
Acrylamide, 99.9%, Thermo Scientific Chemicals
Acrylamide, 99.9%, Thermo Scientific Chemicals
Acrylamide, 99.9%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acrylamide, 99.9%, Thermo Scientific Chemicals
CAS: 79-06-1 | C3H5NO | 71.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALF036545.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAcrylamide
CAS79-06-1
Health Hazard 1H301-H312+H332-H315-H317-H319-H340-H350-H361f-H372
Health Hazard 2GHS H Statement
H301-H340-H350-H372-H361-H312-H332-H315-H319-H317
Health Hazard 3P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c
View more
Acrylamide is primarily used in research laboratories for electrophoresis, chromatography, electron microscopy and in the sewage and wastewater treatment industries. It is used in the production of polymers mainly in the pulp and paper industry and textile treatment industry. The largest use for acrylamide is as an intermediate in the production of organic chemicals and in the synthesis of polyacrylamides.