Lecithin, Refined
Lecithin, Refined
Lecithin, Refined
Thermo Scientific Chemicals
Lecithin, Refined
CAS: 8002-43-5 | C42H80NO8P | 758.075 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF036486.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialLecithin
CAS8002-43-5
Health Hazard 1H373
Health Hazard 3P260-P314-P501c
Recommended StorageAmbient temperatures
View more
Lecithin is used as an emulsifying and stabilizing agent in the food, pharmaceutical, and cosmetic industries. Lecithin is taken as a medicine and is also used in the manufacturing of medicines. Soybean lecithin is useful for its application in liquid ram semen. Fractionation process of sunflower lecithin with absolute ethanol was used for obtaining enriched fractions in certain phospholipids.