Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
CAS: 123-99-9 | C9H16O4 | 188.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALF036308.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAzelaic acid
CAS123-99-9
Melting Point∼102°C
Boiling Point286°C (100mmHg)
Recommended StorageAmbient temperatures
View more
Useful in lubrication and plasticizers. Azelaic acid is used to treat acne.