Straight Wall Nickel Crucible
Straight Wall Nickel Crucible
Straight Wall Nickel Crucible
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
500 mL
20 mL
250 mL
Diameter (Metric) Outer:
View all
Outside: 101mm
Catalog number ALF036088.KT
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
Diameter (Metric) Outer:
Outside: 101mm
Request bulk or custom format
Chemical Identifiers
CAS947533-51-9
IUPAC Name[5-(trifluoromethyl)pyridin-3-yl]boronic acid
Molecular FormulaC6H5BF3NO2
InChI KeySFBQNNGMEKUJAN-UHFFFAOYSA-N
SMILESOB(O)C1=CC(=CN=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=94.0 %