Straight Wall Nickel Crucible
Straight Wall Nickel Crucible
Straight Wall Nickel Crucible
Have Questions?
Change viewbuttonViewtableView
Capacity (Metric):
20 mL
500 mL
250 mL
Diameter (Metric) Outer:
View all
Outside: 33mm
Catalog number ALF035952.KT
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
20 mL
Diameter (Metric) Outer:
Outside: 33mm
Request bulk or custom format
Chemical Identifiers
CAS38861-78-8
IUPAC Name1-[4-(2-methylpropyl)phenyl]ethan-1-one
Molecular FormulaC12H16O
InChI KeyKEAGRYYGYWZVPC-UHFFFAOYSA-N
SMILESCC(C)CC1=CC=C(C=C1)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5140-1.5190 @ 20?C
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to yellow