Hydrochloric acid, 0.5N Standardized Solution, Thermo Scientific Chemicals
Hydrochloric acid, 0.5N Standardized Solution, Thermo Scientific Chemicals
Hydrochloric acid, 0.5N Standardized Solution, Thermo Scientific Chemicals
Hydrochloric acid, 0.5N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrochloric acid, 0.5N Standardized Solution, Thermo Scientific Chemicals
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF035642.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Chemical Identifiers
CAS13682-61-6
IUPAC Namegold(3+) potassium tetrachloride
Molecular FormulaAuCl4K
InChI KeyJMONCQYBAMKVQQ-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[K+].[Au+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Yellow to light orange
Assay>=97.5 %
Hydrochloric acid, 0.5N standardized solution is used for volumetric analysis in laboratories.