Hydrochloric acid, 0.5N Standardized Solution, Thermo Scientific Chemicals
Hydrochloric acid, 0.5N Standardized Solution, Thermo Scientific Chemicals
Hydrochloric acid, 0.5N Standardized Solution, Thermo Scientific Chemicals
Hydrochloric acid, 0.5N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrochloric acid, 0.5N Standardized Solution, Thermo Scientific Chemicals
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number ALF035642.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS2170-03-8
IUPAC Name3-methylideneoxolane-2,5-dione
Molecular FormulaC5H4O3
InChI KeyOFNISBHGPNMTMS-UHFFFAOYSA-N
SMILESC=C1CC(=O)OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)65.0-72.0?C
Hydrochloric acid, 0.5N standardized solution is used for volumetric analysis in laboratories.