Potassium hydroxide, 50% w/v aq. soln., Thermo Scientific Chemicals
Potassium hydroxide, 50% w/v aq. soln., Thermo Scientific Chemicals
Potassium hydroxide, 50% w/v aq. soln., Thermo Scientific Chemicals
Potassium hydroxide, 50% w/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium hydroxide, 50% w/v aq. soln., Thermo Scientific Chemicals
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Catalog NumberQuantity
ALF035621.K21 L
Catalog number ALF035621.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS9005-25-8
IUPAC Name2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyGUBGYTABKSRVRQ-UHFFFAOYNA-N
SMILESOCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Loss on drying=<20 %
Residue after ignition=<1 %
Appearance (Color)White
Infrared spectrumConforms
View more
Potassium hydroxide acts as the precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.