Potassium hydroxide, 10% w/v aq. soln., Thermo Scientific Chemicals
Potassium hydroxide, 10% w/v aq. soln., Thermo Scientific Chemicals
Potassium hydroxide, 10% w/v aq. soln., Thermo Scientific Chemicals
Potassium hydroxide, 10% w/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium hydroxide, 10% w/v aq. soln., Thermo Scientific Chemicals
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF035615.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Chemical Identifiers
CAS99-15-0
IUPAC Name2-acetamido-4-methylpentanoic acid
Molecular FormulaC8H15NO3
InChI KeyWXNXCEHXYPACJF-UHFFFAOYNA-N
SMILESCC(C)CC(NC(C)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)156.0-162.0?C
Appearance (Color)White
Potassium hydroxide acts as the precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.