Ammonium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals
Ammonium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals
Ammonium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals
Ammonium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Catalog NumberQuantity
ALF035614.K21 L
Catalog number ALF035614.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS127-19-5
IUPAC NameN,N-dimethylacetamide
Molecular FormulaC4H9NO
InChI KeyFXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILESCN(C)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (CH3COOH)=<0.1 %
Appearance (Form)Clear liquid
Color scale=<10 APHA
Infrared spectrumConforms
GC>=99.0 %