Sulfuric acid, 6.0N Standardized Solution
Sulfuric acid, 6.0N Standardized Solution
Sulfuric acid, 6.0N Standardized Solution
Sulfuric acid, 6.0N Standardized Solution
Thermo Scientific Chemicals
Sulfuric acid, 6.0N Standardized Solution
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantity
ALF035610.K21 L
Catalog number ALF035610.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
CAS7664-93-9
Quantity1 L
Molecular FormulaH2O4S
MDL NumberMFCD00064589
InChI KeyQAOWNCQODCNURD-UHFFFAOYSA-N