Perchloric acid, 0.1N in Acetic Acid Standardized Solution, Thermo Scientific Chemicals
Perchloric acid, 0.1N in Acetic Acid Standardized Solution, Thermo Scientific Chemicals
Perchloric acid, 0.1N in Acetic Acid Standardized Solution, Thermo Scientific Chemicals
Perchloric acid, 0.1N in Acetic Acid Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perchloric acid, 0.1N in Acetic Acid Standardized Solution, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF035608.K21 L
Catalog number ALF035608.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS89-98-5
IUPAC Name2-chlorobenzaldehyde
Molecular FormulaC7H5ClO
InChI KeyFPYUJUBAXZAQNL-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤2.25%
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.5640-1.5690 @ 20?C
FormLiquid
View more
Perchloric acid, 0.1N in acetic acid standardized solution is used in volumetric analysis. It is useful for its super acid properties and it is also used as an etchant for certain metals.