Hydrochloric acid, 50% v/v aq. soln., Thermo Scientific Chemicals
Hydrochloric acid, 50% v/v aq. soln., Thermo Scientific Chemicals
Hydrochloric acid, 50% v/v aq. soln., Thermo Scientific Chemicals
Hydrochloric acid, 50% v/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrochloric acid, 50% v/v aq. soln., Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF035607.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Chemical Identifiers
CAS9003-01-4
IUPAC NamePoly(acrylic acid)
Molecular Formula(C3H4O2)n
SMILESOC(=O)C(-*)C-*
MDL NumberMFCD00084394
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Heavy metals=<10 ppm (Hg, Pb, As, Sb)
Viscosity(neutralized soln., 0.5% dispersion)
Appearance (Color)White
Loss on drying=<2 %
View more
Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.