Sulfuric acid, 5% v/v aq. soln.
Sulfuric acid, 5% v/v aq. soln.
Sulfuric acid, 5% v/v aq. soln.
Sulfuric acid, 5% v/v aq. soln.
Thermo Scientific Chemicals
Sulfuric acid, 5% v/v aq. soln.
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantity
ALF035606.K21 L
Catalog number ALF035606.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialSulfuric acid
CAS7664-93-9
Concentration5% w/v aq. soln.
Health Hazard 1H290-H315-H318-H335
Health Hazard 2GHS H Statement
H314-H319