Potassium hydrogen iodate, 0.025N Standardized Solution
Potassium hydrogen iodate, 0.025N Standardized Solution
Potassium hydrogen iodate, 0.025N Standardized Solution
Potassium hydrogen iodate, 0.025N Standardized Solution
Thermo Scientific Chemicals
Potassium hydrogen iodate, 0.025N Standardized Solution
CAS: 13455-24-8 | HI2KO6 | 389.909 g/mol
Have Questions?
Catalog NumberQuantity
ALF035600.K21 L
Catalog number ALF035600.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS258279-04-8
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (HPLC)≥98.0%
CommentCounter Ion: Reported
Appearance (Color)White