Magnesium sulfate, anhydrous, 99.5% min, Thermo Scientific Chemicals
Magnesium sulfate, anhydrous, 99.5% min, Thermo Scientific Chemicals
Magnesium sulfate, anhydrous, 99.5% min, Thermo Scientific Chemicals
Magnesium sulfate, anhydrous, 99.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Magnesium sulfate, anhydrous, 99.5% min, Thermo Scientific Chemicals
CAS: 7487-88-9 | MgO4S | 120.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
500 g
2 kg
Catalog number ALF033337.A9
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 kg
Request bulk or custom format
Chemical Identifiers
CAS4551-16-0
IUPAC Nametrimethyl[(trimethylsilyl)tellanyl]silane
Molecular FormulaC6H18Si2Te
InChI KeyVMDCDZDSJKQVBK-UHFFFAOYSA-N
SMILESC[Si](C)(C)[Te][Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear orange-brown
Infrared spectrumConforms
Appearance (Form)Liquid
NMR (1H-NMR)>=97.5 %
Magnesium sulfate is suitable for sensitive molecular biology applications. It used as a component of bath salts and beauty product. It is utilized by the athletes to soothe sore muscles and effective in the removal of splinters. It is commonly used as a desiccant in organic synthesis due to its affinity for water. It acts as a brewing salt in beer production to adjust the ion content of the brewing water as well as enhance enzyme action. It plays a vital role as an antidote for barium chloride poisoning. . It is useful in controlling seizures associated with epilepsy, glomerulonephritis, or hypothyroidism.